accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCl

CAS RN 4981-63-9

IUPAC Name: 1-(4-chlorophenyl)butan-1-one
Synonyms: (4-chlorophenyl) propyl ketone
Molecular Weight (g/mol): 182.65
Molecular Formula: C10H11ClO
InChi Key: XLCJPQYALLFIPW-UHFFFAOYSA-N
SMILES: CCCC(=O)C1=CC=C(Cl)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results