accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 49823-14-5

NN

CAS RN 49823-14-5

IUPAC Name: 1-(2-phenylethyl)-1H-imidazole
Synonyms: 1-(2-phenylethyl)imidazole
Molecular Weight (g/mol): 172.23
Molecular Formula: C11H12N2
InChi Key: SSENHTOBLYRWKU-UHFFFAOYSA-N
SMILES: C(CC1=CC=CC=C1)N1C=CN=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results