CAS RN 50-18-0
IUPAC Name:
2-[bis(2-chloroethyl)amino]-1,3,2λ⁵-oxazaphosphinan-2-one
Synonyms:
cyclophosphamide cyclophosphamide exposure occupation neosar cyclophosphamide, (R)-isomer (r)-isomer cyclophosphamide endoxane endoxana monohydrate, cyclophosphamide cyclophosphamide, (S)-isomer ciclofosfamide
Molecular Weight (g/mol):
261.08
Molecular Formula:
C7H15Cl2N2O2P
InChi Key:
CMSMOCZEIVJLDB-UHFFFAOYNA-N
SMILES:
ClCCN(CCCl)P1(=O)NCCCO1