CAS RN 50-63-5
Chloroquine phosphate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Thermo Scientific Chemicals Chloroquine diphosphate salt, 98%
CAS: 50-63-5 Molecular Formula: C18H32ClN3O8P2 Molecular Weight (g/mol): 515.86 MDL Number: MFCD00069852 InChI Key: QKICWELGRMTQCR-UHFFFAOYNA-N Synonym: chloroquine diphosphate,chloroquine phosphate,aralen phosphate,chingamin phosphate,chloroquine bis phosphate,chingaminum,alermine,h-stadur,aralen diphosphate,miniquine PubChem CID: 64927 IUPAC Name: 4-N-(7-chloroquinolin-4-yl)-1-N,1-N-diethylpentane-1,4-diamine;phosphoric acid SMILES: OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1
Chloroquine diphosphate salt, 98%
CAS: 50-63-5 Molecular Formula: C18H32ClN3O8P2 Molecular Weight (g/mol): 515.86 MDL Number: MFCD00069852 InChI Key: QKICWELGRMTQCR-UHFFFAOYNA-N Synonym: chloroquine diphosphate,chloroquine phosphate,aralen phosphate,chingamin phosphate,chloroquine bis phosphate,chingaminum,alermine,h-stadur,aralen diphosphate,miniquine PubChem CID: 64927 SMILES: OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1
Chloroquine Diphosphate 98.0+%, TCI America™
CAS: 50-63-5 Molecular Formula: C18H32ClN3O8P2 Molecular Weight (g/mol): 515.86 MDL Number: MFCD00069852 InChI Key: QKICWELGRMTQCR-UHFFFAOYNA-N Synonym: chloroquine diphosphate,chloroquine phosphate,aralen phosphate,chingamin phosphate,chloroquine bis phosphate,chingaminum,alermine,h-stadur,aralen diphosphate,miniquine PubChem CID: 64927 IUPAC Name: 7-chloro-N-[5-(diethylamino)pentan-2-yl]quinolin-4-amine; bis(phosphoric acid) SMILES: OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1
R&D Systems™ Chloroquine diphosphate
CAS: 50-63-5 Molecular Formula: C18H32ClN3O8P2
Chloroquine diphosphate salt, 99.6%, MP Biomedicals™
CAS: 50-63-5 Molecular Formula: C18H32ClN3O8P2 Molecular Weight (g/mol): 515.86 MDL Number: MFCD00069852 InChI Key: QKICWELGRMTQCR-UHFFFAOYNA-N Synonym: chloroquine diphosphate,chloroquine phosphate,aralen phosphate,chingamin phosphate,chloroquine bis phosphate,chingaminum,alermine,h-stadur,aralen diphosphate,miniquine PubChem CID: 64927 IUPAC Name: 4-N-(7-chloroquinolin-4-yl)-1-N,1-N-diethylpentane-1,4-diamine;phosphoric acid SMILES: OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1
Chloroquine Diphosphate Salt, MP Biomedicals
CAS: 50-63-5 Molecular Formula: C18H32ClN3O8P2 Molecular Weight (g/mol): 515.86 MDL Number: MFCD00069852 InChI Key: QKICWELGRMTQCR-UHFFFAOYNA-N Synonym: chloroquine diphosphate,chloroquine phosphate,aralen phosphate,chingamin phosphate,chloroquine bis phosphate,chingaminum,alermine,h-stadur,aralen diphosphate,miniquine PubChem CID: 64927 SMILES: OP(O)(O)=O.OP(O)(O)=O.CCN(CC)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1