accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CNNNNCH3H3C

CAS RN 500579-04-4

IUPAC Name: 2-[(3-{[2-(dimethylamino)phenyl]methyl}-2-(pyridin-4-yl)-1,3-diazinan-1-yl)methyl]-N,N-dimethylaniline
Synonyms: 2-[(3-{[2-(dimethylamino)phenyl]methyl}-2-(pyridin-4-yl)-1,3-diazinan-1-yl)methyl]-N,N-dimethylaniline
Molecular Weight (g/mol): 429.61
Molecular Formula: C27H35N5
InChi Key: KVQOGDQTWWCZFX-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=CC=C1CN1CCCN(CC2=CC=CC=C2N(C)C)C1C1=CC=NC=C1
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results