accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOCl

CAS RN 500873-57-4

IUPAC Name: N-benzyl-2-chloro-N-phenylbenzamide
Synonyms: N-benzyl-2-chloro-N-phenylbenzamide
Molecular Weight (g/mol): 321.8
Molecular Formula: C20H16ClNO
InChi Key: OWBLYVPLNRYXED-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1C(=O)N(CC1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results