accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNClCl

CAS RN 5013-77-4

IUPAC Name: [(2,4-dichlorophenyl)methyl](methyl)amine
Synonyms: [(2,4-dichlorophenyl)methyl](methyl)amine
Molecular Weight (g/mol): 190.07
Molecular Formula: C8H9Cl2N
InChi Key: GUJXWKXDISDARD-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(Cl)C=C1Cl
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)