Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-N-methylbenzylamine 98.0+%, TCI America™
SDP

Catalog No. D51475G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51475G 5 g
D51471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D51475G Supplier TCI America Supplier No. D51475G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5013-77-4
Molecular Formula C8H9Cl2N
Molecular Weight (g/mol) 190.067
MDL Number MFCD00045185
InChI Key GUJXWKXDISDARD-UHFFFAOYSA-N
Synonym 2,4-dichlorobenzyl methylamine, n-methyl-2,4-dichlorobenzylamine, benzenemethanamine, 2,4-dichloro-n-methyl, 2,4-dichlorophenyl methyl methyl amine, 2,4-dichloro-n-methylbenzylamine, 1-2,4-dichlorophenyl-n-methylmethanamine, 2,4-dichlorophenyl-n-methylmethanamine, n-2,4-dichlorobenzyl-n-methylamine, 1-2,4-dichlorophenyl-n-methyl-methanamine, 2,4-dichlorobenzyl methyl amine
PubChem CID 485424
IUPAC Name 1-(2,4-dichlorophenyl)-N-methylmethanamine
SMILES CNCC1=C(C=C(C=C1)Cl)Cl

Specifications

Color Yellow
Boiling Point 123°C
Quantity 5 g
UN Number 2735
Formula Weight 190.07
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,4-Dichloro-N-methylbenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.