accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 501435-91-2

HOBOHBrNF

CAS RN 501435-91-2

IUPAC Name: (5-bromo-2-fluoropyridin-3-yl)boronic acid
Synonyms: 5-bromo-2-fluoropyridin-3-ylboronic acid
Molecular Weight (g/mol): 219.8
Molecular Formula: C5H4BBrFNO2
InChi Key: YAXKWQSQZIVTFC-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(Br)=CN=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results