accessibility menu, dialog, popup

UserName

Viewing profile as user:
O

CAS RN 502-72-7

IUPAC Name: cyclopentadecanone
Synonyms: cyclopentadecanone
Molecular Weight (g/mol): 224.39
Molecular Formula: C15H28O
InChi Key: OSOIQJGOYGSIMF-UHFFFAOYSA-N
SMILES: O=C1CCCCCCCCCCCCCC1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)