Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentadecanone 98.0+%, TCI America™
SDP

Catalog No. C142225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C142225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C142225G Supplier TCI America Supplier No. C142225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 502-72-7
Molecular Formula C15H28O
Molecular Weight (g/mol) 224.388
MDL Number MFCD00001266
InChI Key OSOIQJGOYGSIMF-UHFFFAOYSA-N
Synonym exaltone, normuscone, normuscon, unii-t09g246let, exalton, cyclopentadecanon, cylcopentadecanone, cyclopentandecanone, cyclopentadecan-1-one, cyclopentadecanone
PubChem CID 10409
IUPAC Name cyclopentadecanone
SMILES C1CCCCCCCC(=O)CCCCCC1

Specifications

Melting Point 67°C
Color White
Quantity 25 g
Formula Weight 224.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Cyclopentadecanone
EINECSNumber (3)-3307
RTECSNumber GY0900000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.