accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50325-49-0

IUPAC Name: 1-(4-ethenylphenyl)methanamine
Synonyms: 1-(4-ethenylphenyl)methanamine
Molecular Weight (g/mol): 133.19
Molecular Formula: C9H11N
InChi Key: WAVDSLLYAQBITE-UHFFFAOYSA-N
SMILES: NCC1=CC=C(C=C)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results