Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Vinylbenzylamine (stabilized with MEHQ) 92.0+%, TCI America™
SDP

Catalog No. V01015G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V01015G 5 g
V01011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V01015G Supplier TCI America Supplier No. V01015G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50325-49-0
Molecular Formula C9H11N
Molecular Weight (g/mol) 133.194
MDL Number MFCD08704374
InChI Key WAVDSLLYAQBITE-UHFFFAOYSA-N
Synonym 4-(Aminomethyl)styrene
PubChem CID 11073466
IUPAC Name (4-ethenylphenyl)methanamine
SMILES C=CC1=CC=C(C=C1)CN

Specifications

Color Yellow
Boiling Point 60°C
Quantity 5 g
Formula Weight 133.19
Percent Purity ≥92.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Vinylbenzylamine (stabilized with MEHQ)
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.