accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50370-12-2

IUPAC Name: (6R,7R)-7-[(2R)-2-amino-2-(4-hydroxyphenyl)acetamido]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Synonyms: cephadroxil cefadroxil anhydrous cefadroxilo cefadroxilum D-cefadroxil
Molecular Weight (g/mol): 363.39
Molecular Formula: C16H17N3O5S
InChi Key: BOEGTKLJZSQCCD-UEKVPHQBSA-N
SMILES: CC1=C(N2[C@H](SC1)[C@H](NC(=O)[C@H](N)C1=CC=C(O)C=C1)C2=O)C(O)=O
Color 
  • (2)
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results