accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 504-60-9

IUPAC Name: (3E)-penta-1,3-diene
Synonyms: piperylene
Molecular Weight (g/mol): 68.12
Molecular Formula: C5H8
InChi Key: PMJHHCWVYXUKFD-SNAWJCMRSA-N
SMILES: C\C=C\C=C
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)