Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperylene, (Tech.), 60% cis and trans Isomers, Stabilized, Thermo Scientific™

Catalog No. AC207650500
Change view
Click to view available options
Quantity:
50 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC207650500 50 mL
AC207650010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC207650500 Supplier Thermo Scientific Chemicals Supplier No. 207650500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 504-60-9
Molecular Formula C5H8
Molecular Weight (g/mol) 68.119
InChI Key PMJHHCWVYXUKFD-SNAWJCMRSA-N
Synonym 1,3-pentadiene, trans-1,3-pentadiene, piperylene, trans-piperylene, e-1,3-pentadiene, 1-methylbutadiene, 3e-penta-1,3-diene, trans-1-methylbutadiene, 1,3-pentadiene, 3e, trans-penta-1,3-diene
PubChem CID 62204
ChEBI CHEBI:74165
IUPAC Name (3E)-penta-1,3-diene
SMILES CC=CC=C

Specifications

Density 0.68
Infrared Spectrum Authentic
Linear Formula CH3CH=CHCH=CH2
Quantity 50 mL
Beilstein 01, IV, 995
Fieser 01,890
Merck Index 15, 7220
Stabilizer 0.02% max. BHT
Formula Weight 68.11
Percent Purity ≥55%
Grade Technical
Physical Form Liquid
Chemical Name or Material Piperylene, 60% cis and trans isomers
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 207-995-2
RTECSNumber RZ2464000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.