accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3CH3CCH3CH3OH

CAS RN 50428-39-2

IUPAC Name: 2,2,8,8-tetramethylnona-3,6-diyn-5-ol
Synonyms: 2,2,8,8-tetramethylnona-3,6-diyn-5-ol
Molecular Weight (g/mol): 192.3
Molecular Formula: C13H20O
InChi Key: KGZASKNUKKURJQ-UHFFFAOYSA-N
SMILES: CC(C)(C)C#CC(O)C#CC(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results