accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50481-01-1

IUPAC Name: dibromogolduide; tetrabutylazanium
Synonyms: dibromogolduide; tetrabutylammonium ion
Molecular Weight (g/mol): 599.25
Molecular Formula: C16H36AuBr2N
InChi Key: JHKGBGGRJMVGSW-UHFFFAOYSA-L
SMILES: Br[Au-]Br.CCCC[N+](CCCC)(CCCC)CCCC
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results