accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NN

CAS RN 5053-43-0

IUPAC Name: 2-methylpyrimidine
Synonyms: pyrimidine, 2-methyl-
Molecular Weight (g/mol): 94.12
Molecular Formula: C5H6N2
InChi Key: LNJMHEJAYSYZKK-UHFFFAOYSA-N
SMILES: CC1=NC=CC=N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)