Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylpyrimidine, 98%

Catalog No. AC459860050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459860050 5 g
AC459860010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459860050 Supplier Thermo Scientific Chemicals Supplier No. 459860050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 5053-43-0 | C5H6N2 | 94.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5053-43-0
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.11
InChI Key LNJMHEJAYSYZKK-UHFFFAOYSA-N
Synonym pyrimidine, 2-methyl, 2-methyl pyrimidine, pubchem21974, 2-methyl-1,3-diazine, 2-methylpyrimidine, pyrimidine, 2-methyl-6ci,7ci,8ci,9ci
PubChem CID 78748
IUPAC Name 2-methylpyrimidine
SMILES CC1=NC=CC=N1

Specifications

Melting Point -6.0°C
Density 1.0132g/mL
Boiling Point 130.0°C to 132.0°C
Flash Point 37.8°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Specific Gravity 1.0132
Formula Weight 94.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Methylpyrimidine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H StatementFlammable liquid and vapour.Causes skin irritation.Causes serious eye damage.May cause respiratory irritation.
Precautionary Statement GHS P StatementKeep away from heat/sparks/open flames/hot surfaces.- No smoking.IF ON SKIN: Wash with plenty of soap and water.IF IN EYES: Rinse cautiously with water for several minutes.Remove contact lenses,if present and ea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.