accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2SNH

CAS RN 5055-72-1

IUPAC Name: cyclohexylthiourea
Synonyms: thiourea, cyclohexyl- (9CI)
Molecular Weight (g/mol): 158.26
Molecular Formula: C7H14N2S
InChi Key: LEEHHPPLIOFGSC-UHFFFAOYSA-N
SMILES: NC(=S)NC1CCCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results