accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO(Z)

CAS RN 506-17-2

IUPAC Name: (11Z)-octadec-11-enoic acid
Synonyms: cis-vaccenic acid (11E)-octadec-11-enoic acid asclepic acid
Molecular Weight (g/mol): 282.47
Molecular Formula: C18H34O2
InChi Key: UWHZIFQPPBDJPM-FPLPWBNLSA-N
SMILES: CCCCCC\C=C/CCCCCCCCCC(O)=O
Boiling Point 
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)