Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cis-Vaccenic Acid Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022743 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2743 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2743 Supplier MilliporeSigma Supelco Supplier No. 69301100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

cis-Vaccenic acid belongs to a class of unsaturated fatty acids. It is mostly reported to be produced by many species of heterotrophic bacteria, as well as occurring in other marine organisms such as phytoplanktons. It also finds applications as a bacterial biomarker.

TRUSTED_SUSTAINABILITY

Specifications

CAS 506-17-2
Melting Point 14°C to 15°C (literature)
Density 0.887 g/mL (at 25°C (literature))
Boiling Point 150°C/0.03 mmHg (literature)
Flash Point 230°C (Closed Cup)
Molecular Formula C18H34O2
Linear Formula CH3(CH2)5 CH=CH(CH2)9 CO2H
Refractive Index n20/D 1.459 (literature)
Quantity 100 mg
MDL Number MFCD00063187
Synonym cis-11-Octadecenoic acid
Formula Weight 282.46
Percent Purity ≥97% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.