accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 506-30-9

IUPAC Name: icosanoic acid
Synonyms: arachidic acid arachinsaeure arachidinic acid arachate
Molecular Weight (g/mol): 312.54
Molecular Formula: C20H40O2
InChi Key: VKOBVWXKNCXXDE-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (4)
  • (8)
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (7)
Quantity 
  • (4)
  • (2)
  • (5)
  • (3)
Boiling Point 
  • (2)
  • (4)
  • (3)
Physical Form 
  • (2)
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (2)
  • (4)
  • (2)
  • (6)
  • (2)
  • (4)
  • (8)
  • (2)
  • (2)
  • (3)
  • (7)
  • (4)
  • (2)
  • (5)
  • (3)
  • (2)
  • (4)
  • (3)
  • (2)
  • (4)
  • (3)