Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arachidic Acid, Spectrum™ Chemical
SDP

Catalog No. 18620139 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-139 25 g
18-620-136 1 g
18-620-137 5 g
18-620-138 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-620-139 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A389925GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 506-30-9
Molecular Weight (g/mol) 312.53 g/mol

Specifications

Boiling Point 328°C
Quantity 25 g
Percent Purity 99%
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Arachidic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.