Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Eicosanoic acid, 98%

Catalog No. AC117720250
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC117720250 25 g Glass bottle
AC117720010 1 g Glass bottle
AC117720100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC117720250 Supplier Thermo Scientific Chemicals Supplier No. 117720250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 506-30-9 | C20H40O2 | 312.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-30-9
Molecular Formula C20H40O2
Molecular Weight (g/mol) 312.54
MDL Number MFCD00002755
InChI Key VKOBVWXKNCXXDE-UHFFFAOYSA-N
Synonym arachidic acid, eicosanoic acid, arachic acid, n-eicosanoic acid, arachidinic acid, arachidate, eicosanoate, unii-pqb8mjd4rb, pqb8mjd4rb, n-eicosanoate
PubChem CID 10467
ChEBI CHEBI:28822
IUPAC Name icosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCC(O)=O

Specifications

Melting Point 74.0°C to 77.0°C
Color White
Boiling Point 328.0°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3(CH2)18CO2H
Quantity 25 g
Merck Index 15, 753
Solubility Information Solubility in water: practically insoluble. Other solubilities: freely soluble in hot absolute alcohol, benzene,, chloroform and ether
Formula Weight 312.52
Percent Purity 99%
Physical Form Glistening Powder or Flakes
Chemical Name or Material Eicosanoic acid
Show More Show Less
EINECSNumber 208-031-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.