accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHOHOOHOHHOHO

CAS RN 508-52-1

IUPAC Name: 4-[3a,5a,7,9,10-pentahydroxy-9a-(hydroxymethyl)-11a-methyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]-2,5-dihydrofuran-2-one
Synonyms: ouabagenin
Molecular Weight (g/mol): 438.52
Molecular Formula: C23H34O8
InChi Key: BXSABLKMKAINIU-UHFFFAOYNA-N
SMILES: CC12CC(O)C3C(CCC4(O)CC(O)CC(O)C34CO)C1(O)CCC2C1=CC(=O)OC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results