accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOCH3CH3H3CCH3H3CH3CCH3CH3OOOOCH3CH3CH3H3CH3CH3CCH3CH3OOOOCH3CH3CH3H3CCH3H3CCH3

CAS RN 508181-38-2

IUPAC Name: 1,2-bis(3,5,5-trimethylhexyl) cyclohexane-1,2-dicarboxylate; 1,3-bis(3,5,5-trimethylhexyl) cyclohexane-1,3-dicarboxylate; 1,4-bis(3,5,5-trimethylhexyl) cyclohexane-1,4-dicarboxylate
Synonyms: 1,2-bis(3,5,5-trimethylhexyl) cyclohexane-1,2-dicarboxylate; 1,3-bis(3,5,5-trimethylhexyl) cyclohexane-1,3-dicarboxylate; 1,4-bis(3,5,5-trimethylhexyl) cyclohexane-1,4-dicarboxylate
Molecular Weight (g/mol): 1274
Molecular Formula: C78H144O12
InChi Key: QSNOUBLJFWGBSD-UHFFFAOYNA-N
SMILES: CC(CCOC(=O)C1CCC(CC1)C(=O)OCCC(C)CC(C)(C)C)CC(C)(C)C.CC(CCOC(=O)C1CCCC(C1)C(=O)OCCC(C)CC(C)(C)C)CC(C)(C)C.CC(CCOC(=O)C1CCCCC1C(=O)OCCC(C)CC(C)(C)C)CC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results