accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50978-11-5

IUPAC Name: 3,5-diacetamido-2,4,6-triiodobenzoic acid dihydrate
Synonyms: amidotrizoic acid dihydrate
Molecular Weight (g/mol): 649.95
Molecular Formula: C11H13I3N2O6
InChi Key: JHQKUXXJPHSPOL-UHFFFAOYSA-N
SMILES: O.O.CC(=O)NC1=C(I)C(C(O)=O)=C(I)C(NC(C)=O)=C1I