Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diatrizoic acid dihydrate, 98.87%, MP Biomedicals™

Catalog No. ICN19150110 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19150110 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19150110 Supplier MP Biomedicals, Inc Supplier No. 0219150110
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50978-11-5
Molecular Formula C11H13I3N2O6
Molecular Weight (g/mol) 649.946
InChI Key JHQKUXXJPHSPOL-UHFFFAOYSA-N
Synonym diatrizoic acid dihydrate, amidotrizoic acid dihydrate, 3,5-diacetamido-2,4,6-triiodobenzoic acid dihydrate, dsstox_cid_28886, dsstox_rid_83154, dsstox_gsid_48960, 2,4,6-triiodo-3,5-diacetamidobenzoic acid dihydrate, 3,5-bis acetylamino-2,4,6-triiodobenzoic acid dihydrate, 3,5-diacetamido-2,4,6-triiodobenzoic acid-water 1/2, amidotrizoic acid ban:jan
PubChem CID 12598086
ChEBI CHEBI:59733
IUPAC Name 3,5-diacetamido-2,4,6-triiodobenzoic acid;dihydrate
SMILES CC(=O)NC1=C(C(=C(C(=C1I)C(=O)O)I)NC(=O)C)I.O.O

Specifications

Color White
Assay Percent Range 0.9887
Quantity 10 g
Formula Weight 649.95
Physical Form Powder
Chemical Name or Material Diatrizoic acid dihydrate
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.