accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50984-88-8

IUPAC Name: 3,3-bis(4-hydroxy-2,5-dimethylphenyl)-1,3-dihydro-2-benzofuran-1-one
Synonyms: 3,3-bis(4-hydroxy-2,5-dimethylphenyl)-2-benzofuran-1-one
Molecular Weight (g/mol): 374.44
Molecular Formula: C24H22O4
InChi Key: PXCIPOXPHMTCIL-UHFFFAOYSA-N
SMILES: CC1=CC(=C(C)C=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C=C1C
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results