Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Xylenolphthalein, TCI America™
SDP

Catalog No. X00171G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X00171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. X00171G Supplier TCI America Supplier No. X00171G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50984-88-8
Molecular Formula C24H22O4
Molecular Weight (g/mol) 374.436
MDL Number MFCD00042785
InChI Key PXCIPOXPHMTCIL-UHFFFAOYSA-N
PubChem CID 628858
IUPAC Name 3,3-bis(4-hydroxy-2,5-dimethylphenyl)-2-benzofuran-1-one
SMILES CC1=CC(=C(C=C1C2(C3=CC=CC=C3C(=O)O2)C4=CC(=C(C=C4C)O)C)C)O

Specifications

Quantity 1 g
Formula Weight 374.44
Physical Form Crystalline Powder
Chemical Name or Material p-Xylenolphthalein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.