accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51050-59-0

IUPAC Name: 3,4-dichloro-1H-isochromen-1-one
Synonyms: 3,4-dichloroisocoumarin C9H4cl2O2
Molecular Weight (g/mol): 215.03
Molecular Formula: C9H4Cl2O2
InChi Key: SUGXUUGGLDCZKB-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)C2=CC=CC=C2C(=O)O1
Form 
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results