Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dichloroisocoumarin, 98%

Catalog No. AAJ65198MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65198MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ65198MA Supplier Thermo Scientific Chemicals Supplier No. J65198MA
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 51050-59-0 | C9H4Cl2O2 | 215.03 g/mol

Useful in a rapid staining method for identification of macrophages; counts by this method were confirmed by the more complex morphological criteria, by phagocytosis, and by the presence of Fc receptors. It is an inhibitor of Chymotrypsin and Trypsin. ,4 Dichloroisocoumarin also inhibits granzymes A, B, and H, cathepsin G, neutrophil elastase, proteinase and blocks apoptotic internucleosomal DNA cleavage in thymocytes without the involvement of endonucleases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful in a rapid staining method for identification of macrophages; counts by this method were confirmed by the more complex morphological criteria, by phagocytosis, and by the presence of Fc receptors. It is an inhibitor of Chymot
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51050-59-0
Molecular Formula C9H4Cl2O2
Molecular Weight (g/mol) 215.03
MDL Number MFCD00036960
InChI Key SUGXUUGGLDCZKB-UHFFFAOYSA-N
Synonym 3,4-dichloroisocoumarin, 3,4-dcl, 1h-2-benzopyran-1-one,3,4-dichloro, 3,4-dci, 3,4 dichloroisocoumarin, 3,4-dichloro-1h-isochromen-1-one, 3,4-dichloro-1h-2-benzopyran-1-one, 1h-2-benzopyran-1-one, 3,4-dichloro, chembl24983, 3,4-dichloro-2-benzopyran-1-one
PubChem CID 1609
IUPAC Name 3,4-dichloroisochromen-1-one
SMILES ClC1=C(Cl)C2=CC=CC=C2C(=O)O1

Specifications

Color White
Quantity 10 mg
UN Number UN2811
Beilstein 6802625
Solubility Information Soluble in DMF or DMSO at a concentration of 10mM
Formula Weight 215.03
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3,4-Dichloroisocoumarin
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.