accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 51076-95-0

IUPAC Name: 2-chloro-1,1,1-triethoxyethane
Synonyms: 2-chloro-1,1,1-triethoxyethane
Molecular Weight (g/mol): 196.67
Molecular Formula: C8H17ClO3
InChi Key: URFKLQSFBXBOQU-UHFFFAOYSA-N
SMILES: CCOC(CCl)(OCC)OCC
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)