accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN

CAS RN 510775-24-3

IUPAC Name: N-[4-(9-{4-[(naphthalen-1-yl)(phenyl)amino]phenyl}-9H-fluoren-9-yl)phenyl]-N-phenylnaphthalen-1-amine
Synonyms: N-[4-(9-{4-[naphthalen-1-yl(phenyl)amino]phenyl}fluoren-9-yl)phenyl]-N-phenylnaphthalen-1-amine
Molecular Weight (g/mol): 752.96
Molecular Formula: C57H40N2
InChi Key: LKFCORSLOUZJMB-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)N(C1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=C2C=CC=CC2=CC=C1)C1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results