CAS RN 51116-00-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Guanosine 3',5'-cyclic Monophosphate, N2,2'-O-Dibutyryl-, Sodium Salt
Cell-permeable cGMP analog that preferentially activates protein kinase G.