accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NONBrN(R)O(R)OPHOOO(S)(R)OHHN

CAS RN 51116-01-9

IUPAC Name: 9-[(4aR,6R,7R,7aS)-2,7-dihydroxy-2-oxo-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-6-yl]-2-amino-8-bromo-6,9-dihydro-3H-purin-6-one
Synonyms: 9-[(4aR,6R,7R,7aS)-2,7-dihydroxy-2-oxo-tetrahydro-4H-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-6-yl]-2-amino-8-bromo-3H-purin-6-one
Molecular Weight (g/mol): 424.1
Molecular Formula: C10H11BrN5O7P
InChi Key: YUFCOOWNNHGGOD-UMMCILCDSA-N
SMILES: NC1=NC(=O)C2=C(N1)N([C@@H]1O[C@@H]3COP(O)(=O)O[C@H]3[C@H]1O)C(Br)=N2
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results