accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH(R)O(R)HOO(R)O(R)HO(S)OH(R)OH(S)OH(S)OH

CAS RN 512-66-3

IUPAC Name: (2R,3R,4S,5R)-2-{[(2R,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}oxane-3,4,5-triol
Synonyms: (2R,3R,4S,5R)-2-{[(2R,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}oxane-3,4,5-triol
Molecular Weight (g/mol): 312.27
Molecular Formula: C11H20O10
InChi Key: DRYHSZUFKNRFCT-OPNUYLJJSA-N
SMILES: OC[C@H]1O[C@](CO)(O[C@H]2OC[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results