accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH2NOOH3C

CAS RN 5121-34-6

IUPAC Name: methyl 2-amino-3-methoxybenzoate
Synonyms: methyl 2-amino-3-methoxybenzoate
Molecular Weight (g/mol): 181.19
Molecular Formula: C9H11NO3
InChi Key: YJEZEMGLLFLMDF-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N)C(OC)=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results