Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-Amino-3-methoxybenzoate 98.0+%, TCI America™
SDP

Catalog No. M28225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28225G 5 g
M28221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28225G Supplier TCI America Supplier No. M28225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5121-34-6
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.191
MDL Number MFCD11113109
InChI Key YJEZEMGLLFLMDF-UHFFFAOYSA-N
Synonym 2-Amino-3-methoxybenzoic Acid Methyl Ester, Methyl 2-Amino-m-anisate, 2-Amino-m-anisic Acid Methyl Ester
PubChem CID 602320
IUPAC Name methyl 2-amino-3-methoxybenzoate
SMILES COC1=CC=CC(=C1N)C(=O)OC

Specifications

Melting Point 48°C
Boiling Point 117°C
Quantity 5 g
Formula Weight 181.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-Amino-3-methoxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.