accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 513-42-8

IUPAC Name: 2-methylprop-2-en-1-ol
Synonyms: methallyl alcohol
Molecular Weight (g/mol): 72.11
Molecular Formula: C4H8O
InChi Key: BYDRTKVGBRTTIT-UHFFFAOYSA-N
SMILES: CC(=C)CO
Boiling Point 
  • (3)
  • (6)
Molecular Weight (g/mol) 
  • (4)
  • (5)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (6)
  • (4)
  • (5)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)