Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-2-propen-1-ol, 98%

Catalog No. AC150295000
Change view
Click to view available options
Quantity:
10 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC150295000 500 mL
AC150290100 10 mL
AC150291000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC150295000 Supplier Thermo Scientific Chemicals Supplier No. 150295000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 513-42-8 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-42-8
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00004737
InChI Key BYDRTKVGBRTTIT-UHFFFAOYSA-N
Synonym methallyl alcohol, 2-methyl-2-propen-1-ol, 2-methylallyl alcohol, isopropenyl carbinol, 2-propen-1-ol, 2-methyl, beta-methallyl alcohol, methacryl alcohol, 3-hydroxy-2-methylpropene, beta-methylallyl alcohol, 2-methyl-2-propene-1-ol
PubChem CID 10557
IUPAC Name 2-methylprop-2-en-1-ol
SMILES CC(=C)CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -50°C
Density 0.8500g/mL
Boiling Point 113°C to 115°C
Flash Point 33°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4250 to 1.427
Linear Formula H2C=C(CH3)CH2OH
Quantity 500 mL
Beilstein 01, 443
Specific Gravity 0.85
Solubility Information Solubility in water: 11 %. Other solubilities: soluble in alcohols,esters,ketones and,hydrocarbons
Viscosity 2.24 mPa.s (20°C)
Formula Weight 72.11
Percent Purity 98%
Chemical Name or Material 2-Methyl-2-propen-1-ol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-161-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.