Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-2-propen-1-ol, 98%

Catalog No. AAH6050222
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6050222 100 g
AAH6050206 5 g
AAH6050214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6050222 Supplier Thermo Scientific Chemicals Supplier No. H6050222
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 513-42-8 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-42-8
Molecular Formula C4H8O
MDL Number MFCD00004737
InChI Key BYDRTKVGBRTTIT-UHFFFAOYSA-N
PubChem CID 10557
IUPAC Name 2-methylprop-2-en-1-ol
SMILES CC(=C)CO

Specifications

Density 0.857
Boiling Point 113°C to 115°C
Flash Point 32°C
Refractive Index 1.426
Linear Formula CH2=C(CH3)CH2OH
Quantity 100 g
UN Number UN2614
Beilstein 969226
Synonym methallyl alcohol, 2-methyl-2-propen-1-ol
Molecular Weight (g/mol) 72.11
Formula Weight 72.11
Percent Purity 98%
Chemical Name or Material 2-Methyl-2-propen-1-ol
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement

H226-H315-H319-H335

Flammable liquid and vapor.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: METHALLYL ALCOHOL
EINECSNumber 208-161-0
RTECSNumber UD5250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.