accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5137-69-9

IUPAC Name: undecylphosphonic acid
Synonyms: undecylphosphonic acid
Molecular Weight (g/mol): 236.29
Molecular Formula: C11H25O3P
InChi Key: GKIQHTGBORJXKZ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCP(O)(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results