accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClHNOH2C

CAS RN 51479-73-3

IUPAC Name: prop-2-en-1-yl 2,2,2-trichloroethanimidate
Synonyms: prop-2-en-1-yl 2,2,2-trichloroethanimidate
Molecular Weight (g/mol): 202.46
Molecular Formula: C5H6Cl3NO
InChi Key: UJYZRNWTLPBNOR-UHFFFAOYSA-N
SMILES: ClC(Cl)(Cl)C(=N)OCC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results