CAS RN 51481-10-8
IUPAC Name:
(1'R,2S,2'R,3'S,7'R,9'R,10'R)-3',10'-dihydroxy-2'-(hydroxymethyl)-1',5'-dimethyl-8'-oxaspiro[oxirane-2,12'-tricyclo[7.2.1.0²,⁷]dodecan]-5'-en-4'-one
Synonyms:
vomitoxin
Molecular Weight (g/mol):
296.32
Molecular Formula:
C15H20O6
InChi Key:
LINOMUASTDIRTM-QGRHZQQGSA-N
SMILES:
CC1=C[C@H]2O[C@@H]3[C@H](O)C[C@@](C)([C@]33CO3)[C@@]2(CO)[C@H](O)C1=O