Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Deoxynivalenol
SDP

Catalog No. 39761 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-761 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 39-761 Supplier Tocris Bioscience™ Supplier No. 3976/1

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mycotoxin; potent protein synthesis inhibitor

Tricothecene mycotoxin and potent protein synthesis inhibitor. Exhibits cytotoxic activity in vivo via the ribotoxic stress response. Induces p38-mediated gene expression and apoptosis in leukocytes; activity results in systemic expression of interleukin-6 (IL-6) and other proinflammatory cytokines. Also induces migration of NF-κB into the nucleus.

Chemical Identifiers

CAS 51481-10-8
Molecular Formula C15H20O6

Specifications

Quantity 1 mg
Formula Weight 296.32
Purity 98%
Molecular Weight (g/mol) 296.32
Chemical Name or Material Deoxynivalenol
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.