accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaOSOO

CAS RN 515-42-4

IUPAC Name: sodium benzenesulfonate
Synonyms: sodium benzenesulfonate
Molecular Weight (g/mol): 180.15
Molecular Formula: C6H5NaO3S
InChi Key: MZSDGDXXBZSFTG-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (6)