Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Benzenesulfonate 96.0+%, TCI America™
SDP

Catalog No. B0035500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0035500G 500 g
B003525G 25 g
B0035100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0035500G Supplier TCI America Supplier No. B0035500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 515-42-4
Molecular Formula C6H5NaO3S
Molecular Weight (g/mol) 180.153
MDL Number MFCD00065179
InChI Key MZSDGDXXBZSFTG-UHFFFAOYSA-M
Synonym sodium benzenesulfonate, benzenesulfonic acid sodium salt, benzenesulfonic acid, sodium salt, dispersant gn, sodium phenylsulfonate, sodium benzenesulphonate, sodium benzene sulfonate, sodium benzene monosulfate, unii-k5rm14azhx, sodium benzene monosulfonate
PubChem CID 517327
IUPAC Name sodium;benzenesulfonate
SMILES C1=CC=C(C=C1)S(=O)(=O)[O-].[Na+]

Specifications

Color White
Quantity 500 g
Formula Weight 180.15
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Benzenesulfonate
EINECSNumber (3)-1928
RTECSNumber DB7350000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.